Counting chambers, Neubauer
供应商: MARIENFELD
0640111 0640110 0640130 0640131
MARI0640111EA 0 CNY
MARI0640111 MARI0640110 MARI0640130 MARI0640131
Counting chambers, Neubauer
载玻片和盖玻片 细胞计数板
These counting chambers are designed for counting leucocytes and erythrocytes.
  • Double net ruling
  • Two counting grids with one division
Certifications: IVD 98/79/EC
Delivery information: Supplied with two cover glasses.