硫酸锌七水合物, granular USP, FCC, Macron Fine Chemicals™

供应商: Avantor Performance Materials
Danger

MACR8872-07EA 0 CNY
MACR8872-07 MACR8872-24 MACR8872-20
硫酸锌七水合物, granular USP, FCC, Macron Fine Chemicals™
硫酸锌七水合物
分子式: ZnSO₄·7H₂O
分子量: 287.54 g/mol
溶点: 100
密度: 1.97 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00149894
CAS号: 7446-20-0
EINECS: 231-793-3
UN号: 3077
ADR: 9 III
Merck Index: 12,10293

现在订购

产品技术规格

Meets U.S.P Requirements
Food GMP Manufactured Product
GMP Manufactured Product
Meets F.C.C. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
Assay ((as ZnSO₄)) 55.6 - 61.0 %
Assay (ZnSO₄·7H₂O) 99.0 - 108.7 %
Identification A Passes Test
Identification B Passes Test
Appearance Passes Test
Acidity Passes Test
FCC - Alkalies and Alkaline Earths ≤0.5 %
USP - Alkalies and Alkaline Earths ≤0.9 %
Arsenic (As) ≤3 ppm
FCC - Iron (Fe) ≤0.003 %
FCC - Lead (Pb)(FCC) ≤4 mg/kg
USP - Lead (Pb) ≤0.002 %
FCC - Cadmium (Cd) ≤2 mg/kg
FCC - Mercury (Hg) ≤5 ppm
FCC - Selenium (Se) ≤0.003 %
Solubility Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR