尿素 98.0-102% USP, Macron Fine Chemicals™

供应商: Avantor Performance Materials

Synonyms: 碳酰胺, 碳酰二胺

MACR8642-30EA 0 CNY
MACR8642-30 MACR8642-12 MACR8642-88
尿素 98.0-102% USP, Macron Fine Chemicals™
尿素
分子式: NH₂CONH₂
分子量: 60.06 g/mol
沸点: 197 °C (1013 hPa)
溶点: 132.7 °C
密度: 1.34 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00008022
CAS号: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

现在订购

产品技术规格

GMP Manufactured Product
Meets U.S.P Requirements
Preserve in tight containers. Store at 25°C, excursions permitted 15-30°C.
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Alcohol Insoluble Matter ≤0.04 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Residue on Ignition ≤0.1 %
USP - Organic impurities: Urea related compound A ≤0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤0.1 %
USP - Organic impurities: Total impurities ≤2.0 %
Appearance (colorless to white powder, granules, or small pellets)
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR