乙二胺四乙酸 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™

供应商: Avantor Performance Materials
Warning

Synonyms: 依地酸, Ethylenedinitrilotetraacetic acid, EDTA, 亚乙基二次氮基四乙酸

Management of Change (MOC) category = N

MACR2580-12EA 499 CNY
MACR2580-12
乙二胺四乙酸 99.4-100.6%, powder, AR® ACS, Macron Fine Chemicals™
乙二胺四乙酸

Powder.

分子式: C₁₀H₁₆N₂O₈
分子量: 292.25 g/mol
沸点: 614 °C (1013 hPa)
溶点: 220 °C
密度: 0.86 g/cm³ (20 °C)
闪点: >100 °C
存储条件: Ambient
MDL编号: MFCD00003541
CAS号: 60-00-4
EINECS: 200-449-4
REACH: 01-2119486399-18
Merck Index: 13,03546
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

现在订购

产品技术规格

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₁₀H₁₆N₂O₈) 99.4 - 100.6 %
ACS - Calcium (Ca) ≤0.001 %
ACS - Heavy Metals (as Pb) ≤0.001 %
ACS - Insoluble in Dilute NH₄OH ≤0.005 %
ACS - Iron (Fe) ≤0.005 %
ACS - Magnesium (Mg) ≤5 ppm
ACS - Nitriloacetic Acid ≤0.1 %
ACS - Residue after Ignition ≤0.2 %

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR