碘化钾 ≥99.0%, AR® ACS, free-flowing, Macron Fine Chemicals™

供应商: Avantor Performance Materials
Danger

Synonyms: Dipotassium diiodide, Kali iodide

Management of Change (MOC) category = N

MACR1123-20EA 0 CNY
MACR1123-20 MACR1123-02 MACR1123-06 MACR1123-12 MACR1123-24
碘化钾 ≥99.0%, AR® ACS, free-flowing, Macron Fine Chemicals™
碘化钾
分子式: Kl
分子量: 166 g/mol
沸点: 1330 °C (1013 hPa)
溶点: 681 °C
密度: 3.13 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011405
CAS号: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

现在订购

产品技术规格

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use ≥99.0%
ACS - Assay (KI) ≥99.0 %
ACS - Appearance (white, crystalline granules) Passes Test
ACS - Barium (Ba) ≤0.002 %
ACS - Calcium (Ca) ≤0.002 %
ACS - Chloride and Bromide (as Cl) ≤0.01 %
ACS - Heavy Metals (as Pb) ≤0.0005 %
ACS - Insoluble Matter ≤0.005 %
ACS - Iodate (IO₃) ≤3 ppm
ACS - Iron (Fe) ≤0.0003 %
ACS - Loss on Drying at 150°C ≤0.2 %
ACS - Magnesium (Mg) ≤0.001 %
ACS - pH of 5% Solution at 25°C 6.0 - 9.2
ACS - Phosphate (PO₄) ≤0.001 %
ACS - Sodium (Na) ≤0.005 %
ACS - Sulfate (SO₄) ≤0.005 %
Free Alkali, For Information Only For Information Only
Total Alkali, For Information Only

规格

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR