Filter funnels
供应商: LENZ LABORGLAS
07195428 07195379 07195328 07195334 07195340 07195358 07195434 07195440 07195458 07195258 07195128 07195134 07195140 07195158 07195170 07195179 07195228 07195234 07195240 07195270 07195279 07195370 07195470 07195479
LENZ07195428EA 0 CNY
LENZ07195428 LENZ07195379 LENZ07195328 LENZ07195334 LENZ07195340 LENZ07195358 LENZ07195434 LENZ07195440 LENZ07195458 LENZ07195258 LENZ07195128 LENZ07195134 LENZ07195140 LENZ07195158 LENZ07195170 LENZ07195179 LENZ07195228 LENZ07195234 LENZ07195240 LENZ07195270 LENZ07195279 LENZ07195370 LENZ07195470 LENZ07195479
Filter funnels
漏斗 过滤漏斗 Filter Funnels
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.