Beakers, low form
供应商: LENZ LABORGLAS
07001010 07001022 07001028 07001037 07001043 07001049 07001055 07001061 07001067 07001070 07001073 07001076 07001082 07001088
LENZ07001010EA 0 CNY
LENZ07001010 LENZ07001022 LENZ07001028 LENZ07001037 LENZ07001043 LENZ07001049 LENZ07001055 LENZ07001061 LENZ07001067 LENZ07001070 LENZ07001073 LENZ07001076 LENZ07001082 LENZ07001088
Beakers, low form
烧杯
DURAN®, borosilicate glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Graduated for approximate reading of contents and labelling area (exept 5 ml beaker, not graduated).
Certifications: DIN 12331 and ISO 3819