Flat flange lids with 3 necks
供应商: LENZ LABORGLAS
06103444 06103464 06103564 06103244 06103141 06103241 06103264 06103544 06103111
LENZ06103444EA 0 CNY
LENZ06103444 LENZ06103464 LENZ06103564 LENZ06103244 LENZ06103141 LENZ06103241 LENZ06103264 LENZ06103544 LENZ06103111
Flat flange lids with 3 necks
盖子 Reaction Vessel Lids
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat laboratory flange (DN). Straight 29/32 standard ground middle neck, and two standard ground side necks.