Drying tubes, straight
供应商: LENZ LABORGLAS
05330013
LENZ05330013EA 0 CNY
LENZ05330013
Drying tubes, straight
管子 干燥管
DURAN®, borosilicate glass, clear. One bulb and glass tubing connection.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes