Adapters with stopcock and connector
供应商: LENZ LABORGLAS
05097001 05097002 05097003 05097004 05098001 05098002 05098003 05098004
LENZ05097001EA 0 CNY
LENZ05097001 LENZ05097002 LENZ05097003 LENZ05097004 LENZ05098001 LENZ05098002 LENZ05098003 LENZ05098004
Adapters with stopcock and connector
适配器 带连接管接头的适配器
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With standard ground cone, standard ground stopcock with solid glass key and a straight or right angled tubing connector. The tubing connector has an external diameter of 8 mm.