Conical separating funnels
供应商: LENZ LABORGLAS
04000028 04000037 04000049 04000058 04000070 04000073 04002073 04004170 04002028 04002037 04002049 04002058 04002070 04004128 04004137 04004149 04004158 04004173 04006128 04006137 04006149 04006158 04006170 04006173
LENZ04000028EA 0 CNY
LENZ04000028 LENZ04000037 LENZ04000049 LENZ04000058 LENZ04000070 LENZ04000073 LENZ04002073 LENZ04004170 LENZ04002028 LENZ04002037 LENZ04002049 LENZ04002058 LENZ04002070 LENZ04004128 LENZ04004137 LENZ04004149 LENZ04004158 LENZ04004173 LENZ04006128 LENZ04006137 LENZ04006149 LENZ04006158 LENZ04006170 LENZ04006173
Conical separating funnels
漏斗 分液漏斗
DURAN®, borosilicate glass, with NS-PE stopper.
With socket and plain stem. Available with interchangeable solid glass key or alternatively PTFE key for grease-free performance. Both with or without graduations.