Flasks, Erlenmeyer, with standard ground joint
供应商: LENZ LABORGLAS
03022428 03022437 03022446 03022449 03022452 03022458 03022470 03022473 03022928 03022937 03022946 03022949 03022952 03022958 03022970 03022973 03022943
LENZ03022428EA 0 CNY
LENZ03022428 LENZ03022437 LENZ03022446 LENZ03022449 LENZ03022452 LENZ03022458 LENZ03022470 LENZ03022473 LENZ03022928 LENZ03022937 LENZ03022946 LENZ03022949 LENZ03022952 LENZ03022958 LENZ03022970 LENZ03022973 LENZ03022943
Flasks, Erlenmeyer, with standard ground joint
烧瓶 三角烧瓶
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.
DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)
Certifications: DIN 12387 (except Cat. No. 201-1114, 201-1122, 201-0318, 201-1116 and 271-1126)