Glass stoppers, standard taper, hexagonal, hollow
供应商: LENZ LABORGLAS
01400007 01400010 01400012 01400014 01400019 01400024 01400029 01400034 01400045 01400345 01400307 01400324 01400314 01400312 01400329 01400310 01400319 01400334
LENZ01400007EA 0 CNY
LENZ01400007 LENZ01400010 LENZ01400012 LENZ01400014 LENZ01400019 LENZ01400024 LENZ01400029 LENZ01400034 LENZ01400045 LENZ01400345 LENZ01400307 LENZ01400324 LENZ01400314 LENZ01400312 LENZ01400329 LENZ01400310 LENZ01400319 LENZ01400334
Glass stoppers, standard taper, hexagonal, hollow
塞子 玻璃塞
These standard taper hollow stoppers are made from borosilicate glass 3.3 and have a hexagonal head.
  • Excellent chemical resistance
  • High thermal resistance, minimal heat expansion results in high resistance to temperature changes
  • Hexagonal head allows easy release and removal of the stopper
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus.
Certifications: DIN 12252
订购须知 Available with flat or pointed bottom.