Centrifugal concentrators, complete systems and system packages, CentriVap®
供应商: LABCONCO
7982011
7983014
7982035
7983035
7989601
7989635
7315062
7315060
7315061
7315063
7995811
7315070
LBCP7982011EA
248880.43
CNY
LBCP7982011
LBCP7983014
LBCP7982035
LBCP7983035
LBCP7989601
LBCP7989635
LBCP7315062
LBCP7315060
LBCP7315061
LBCP7315063
LBCP7995811
LBCP7315070
Centrifugal concentrators, complete systems and system packages, CentriVap®
蒸发器
离心蒸发器
规格
型号 | CentriVap® | CentriVap® | |
Environmentally Preferable | |||
报警器 | Audible alarm indicates end of cycle | Audible alarm indicates end of cycle | |
内存 | 1 - 9 programs | 1 - 9 programs | |
循环时间 | 1 - 999 min | 1 - 999 min | |
加热 | From Off to HI (+100 °C) in increments of 1 degree | From Off to HI (+100 °C) in increments of 1 degree | |
加热时间 | 1 - 999 min | 1 - 999 min |