Accessories for FreeZone® freeze drying systems
供应商: LABCONCO
7522300 7540000 7540200 7540300 7540400 7540600 7540800 7540900 7541000 7545200 7545800 7547200 7547600 7550900 7551000 7568000 7575010 7575210 7762700 7645900 7556400 7557000 7557200 7557400 7557600 7557800 7557900 7558000 7559000 7559200 7559400 7568200 7568400 7568600 7570610 7570810 7571010 7571210 7572710 7573010 7573210 7573610 7574010 7574210 7576010 7576210 7577010 7577110 7578000 7578100 7578500 7394800 7442600 7442700 7509600 7538400 7541100 7541200 7542000 7542200 7542300 7542400 7542600 7542700 7542800 7542900 7543000 7543200 7544000 7544200 7544400 7544810 7545000 7545400 7545600 7546000 7547000 7547400 7550000 7550200 7550400 7550600 7550800 7552000 7552200 7552400 7554000 7554200 7554400 7554600 7554800 7554900 7555000 7556000 7556200 7762800 7772100 7772600 7728000 7426400 7573810 7573410 7439903 7543800 7593401 7386800 7569000 7205100 7317100
LBCP7522300EA 66395.81 CNY
LBCP7522300 LBCP7540000 LBCP7540200 LBCP7540300 LBCP7540400 LBCP7540600 LBCP7540800 LBCP7540900 LBCP7541000 LBCP7545200 LBCP7545800 LBCP7547200 LBCP7547600 LBCP7550900 LBCP7551000 LBCP7568000 LBCP7575010 LBCP7575210 LBCP7762700 LBCP7645900 LBCP7556400 LBCP7557000 LBCP7557200 LBCP7557400 LBCP7557600 LBCP7557800 LBCP7557900 LBCP7558000 LBCP7559000 LBCP7559200 LBCP7559400 LBCP7568200 LBCP7568400 LBCP7568600 LBCP7570610 LBCP7570810 LBCP7571010 LBCP7571210 LBCP7572710 LBCP7573010 LBCP7573210 LBCP7573610 LBCP7574010 LBCP7574210 LBCP7576010 LBCP7576210 LBCP7577010 LBCP7577110 LBCP7578000 LBCP7578100 LBCP7578500 LBCP7394800 LBCP7442600 LBCP7442700 LBCP7509600 LBCP7538400 LBCP7541100 LBCP7541200 LBCP7542000 LBCP7542200 LBCP7542300 LBCP7542400 LBCP7542600 LBCP7542700 LBCP7542800 LBCP7542900 LBCP7543000 LBCP7543200 LBCP7544000 LBCP7544200 LBCP7544400 LBCP7544810 LBCP7545000 LBCP7545400 LBCP7545600 LBCP7546000 LBCP7547000 LBCP7547400 LBCP7550000 LBCP7550200 LBCP7550400 LBCP7550600 LBCP7550800 LBCP7552000 LBCP7552200 LBCP7552400 LBCP7554000 LBCP7554200 LBCP7554400 LBCP7554600 LBCP7554800 LBCP7554900 LBCP7555000 LBCP7556000 LBCP7556200 LBCP7762800 LBCP7772100 LBCP7772600 LBCP7728000 LBCP7426400 LBCP7573810 LBCP7573410 LBCP7439903 LBCP7543800 LBCP7593401 LBCP7386800 LBCP7569000 LBCP7205100 LBCP7317100
Accessories for FreeZone® freeze drying systems
冷冻干燥器
Fast-Freeze® flasks for FreeZone® freeze dry systems are designed to be easier to handle, faster to load, and more convenient to use in the laboratory for freeze drying.
  • Compatible with all major brands of freeze drying equipment, including labconco and Virtis™
Fast-Freeze® flasks simplify lyophilization procedures with a wide-mouth design that allows easy access to samples.
Flat bottom, high-strength borosilicate glass flask withstands high heat and deep vacuum. Uniform glass walls maximize heat transfer while maintaining structural strength. Wide-More Product Information
现在订购
规格
Environmentally Preferable Sustainable Materials,Sustainable Packaging Sustainable Packaging Sustainable Materials