Filtered balance systems, XPert™
供应商: LABCONCO
3950221 3950222 3950321 3950322 3950421 3950622 3950420 3950422 3950320 3950220 3950522
LBCP3950221EA 145173.33 CNY
LBCP3950221 LBCP3950222 LBCP3950321 LBCP3950322 LBCP3950421 LBCP3950622 LBCP3950420 LBCP3950422 LBCP3950320 LBCP3950220 LBCP3950522
Filtered balance systems, XPert™
通风柜 萃取柜
These portable systems provide user protection by keeping powders and particulates contained during weighing procedures. The built-in blower and HEPA filter are mounted in the upper plenum to save space, and the enclosures have low installation costs and require no outside ducting. Systems also provide quiet operation as low as 45 dBA, depending on face velocity.
  • The deep interior easily accommodates large micro and analytical balances
  • The Clean-Sweep™ airfoil allows air to sweep the work surface for maximum containment
  • The perforated rear baffle promotes
More Product Information