F1-ClipTip™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
供应商: Thermo Fisher Scientific
4641180N 4641190N 4641200N 4641210N 4641220N 4641230N 4641310N 4641320N
LABS4641180NEA 0 CNY
LABS4641180N LABS4641190N LABS4641200N LABS4641210N LABS4641220N LABS4641230N LABS4641310N LABS4641320N
F1-ClipTip™ Single-Channel Pipettes, Mechanical, Fixed/Variable Volume
移液管和移液器 移液器
The Thermo Scientific™ F1-ClipTip™ pipette features interlocking technology to ensure secure tip attachment. The pipettes work exclusively with Thermo Scientific ClipTip pipette tips, providing a unique tip interface that locks the tip in place, and ensures a complete seal with minimal tip application and ejection forces. Tips are locked firmly in place and will not fall off.
  • Silent and lighter volume adjustment mechanism with over 50% lower forces compared to earlier models
  • Improved grip on the volume adjustment knob ensures effortless and precise
More Product Information