Flange lids with 4 necks, SP Wilmad-LabGlass
供应商: LAB GLASS WILMAD
LG-8050-104
LABGLG-8050-104EA 0 CNY
LABGLG-8050-104
Flange lids with 4 necks, SP Wilmad-LabGlass
盖子 Reaction Vessel Lids
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Conical flange has four necks with one thermometer neck. Base of lid is finely ground to assure a vacuum tight seal.