Safety knives, Fish 2000/3000
供应商: THE SAFETY KNIFE
FISH3000MBLUE FISH3000ORANGE FISH2000MBLUE FISH2000BLUE
KNIFFISH3000MBLUEEA 0 CNY
KNIFFISH3000MBLUE KNIFFISH3000ORANGE KNIFFISH2000MBLUE KNIFFISH2000BLUE
Safety knives, Fish 2000/3000
刀和刀具
Carbon steel or stainless steel blade, ABS body.
  • Disposable
  • Lanyard point
  • Available with manual or auto-retractable hook blade at the tail
These disposable knives are based on the original fish design with the added feature of the 'opposing slopes' at the mouth exit for a more efficient cut and a manual or an an auto-retractable hook blade at the tail. Either moulded in a metal detectable plastic with stainless steel blades or in standard ABS plastic with carbon steel blades. All versions are ideal for cutting strapping/banding, stretch/bubble wrap,More Product Information