Media/storage bottles, graduated, with screw cap, KIMBLE® KIMAX®
供应商: KIMBLEKONTES
14395-100 14395-5000
KIML14395-100EA 0 CNY
KIML14395-100 KIML14395-5000
Media/storage bottles, graduated, with screw cap, KIMBLE® KIMAX®
玻璃瓶
General-purpose bottles feature white enamel graduations and marking spots. Wide, 30 mm int. Ø openings are designed for easy filling. Replaceable, drip-free, clear polypropylene pour ring is incorporated into the bottle design. Bottles are supplied with linerless GL 45 polypropylene caps that are autoclavable to 140 °C (284 °F). Optional high-temperature caps and pour rings are autoclavable to 180 °C (356 °F).