Chemical resistant gloves, JACKSON SAFETY* G80 NITRILE*
供应商: KIMBERLY CLARK
94446 94447 94448
KIMB94446EA 0 CNY
KIMB94446 KIMB94447 KIMB94448
Chemical resistant gloves, JACKSON SAFETY* G80 NITRILE*
手套
JACKSON SAFETY* G80 NITRILE* gloves provide protection against acids, caustics, bases, solvents and other aqueous solutions.
  • Textured fingertips and palm for enhanced grip
  • Improved, more consistent cotton flock lining for added comfort
  • Approved for food contact in the US, Canada and Europe
Thanks to the modified nitrile formulation the hand-specific gloves feature even more durability and enhanced chemical resistance, thus being suitable for chemical processing and oil refining, automotive assembly and painting, machining operations using cutting oilMore Product Information