Cleanroom wipes, Kimtech™ Pure W4
供应商: KIMBERLY CLARK
33330
KIMB33330EA 0 CNY
KIMB33330
Cleanroom wipes, Kimtech™ Pure W4
擦拭巾 洁净室擦拭巾 Dry Cleanroom Wipes
These critical task wipes are made of 100% PP and acid, base and solvent-resistant.
  • Thermally bonded with no glues or binders
  • Straight, clean, knife-cut edges minimise lint
  • Meet the U.S. FDA requirements for food packaging materials
Ideal for use in pharmaceutical, high-tech, biotechnology and medical-device industries.
Suitable for use in Class 10, ISO 4, M2.5 cleanrooms.
Colour: White
Packaging: Double-bagged in anti-stat packaging.