Beakers, low or tall form
供应商: KAVALIER GLASS
632417010952 632417010956 632417012250 632417012940 632417012950
KAVA632417010952EA 0 CNY
KAVA632417010952 KAVA632417010956 KAVA632417012250 KAVA632417012940 KAVA632417012950
Beakers, low or tall form
烧杯
Borosilicate glass, grade 3,3.
  • Excellent chemical resistance
  • High temperature resistance
  • Autoclavable
Ideal for general laboratory use. With spout and white graduations.
Certifications: DIN 12331 (exept 25 ml/5000 ml tall form and 10000 ml low form supplementary to DIN). ISO 3819.