Bottles, wide neck, with screw cap, series 303
供应商: KAUTEX
303-783855 303-783856 303-783859 303-783852
KAUT303-783855EA 0 CNY
KAUT303-783855 KAUT303-783856 KAUT303-783859 KAUT303-783852
Bottles, wide neck, with screw cap, series 303
塑料瓶
Polypropylene, round, natural colour.
  • Autoclavable up to 121 °C
  • Hot filled products, pastes, granulates
  • Leakproof
  • Halal certified
Suitable for food.
Note: Unscrew the cap and set on the bottle before autoclaving.
Caps are only suitable for bottles 215-9044 to 215-9047.