Pipette tray
供应商: KARTELL
996
KART996EA 0 CNY
KART996
Pipette tray
盘子/皿/盆 吸管托盘
PVC.
  • Very useful workstation holder for items other than pipettes
  • For 7 pipettes diameter <20 mm or for 16 pipettes diameter <10 mm
  • For pipettes of all lengths
W×D×H: 283×216×40 mm