Magnetic stirring bars, with pivot ring
供应商: KARTELL
788 787 786
KART788EA 0 CNY
KART788 KART787 KART786
Magnetic stirring bars, with pivot ring
搅拌器 磁力搅拌子
PTFE coated suitable for use in containers with irregular bottom.
  • Angled surfaces create high turbulence even at low speed
  • Pivot ring reduces the contact surface and fosters optimal stir bar positioning
  • Suitable for foodstuff