Magnetic stirring bars, PTFE coated, crosshead
供应商: KARTELL
771 772 773
KART771EA 0 CNY
KART771 KART772 KART773
Magnetic stirring bars, PTFE coated, crosshead
搅拌器 磁力搅拌子
Cross shaped magnetic stirring bars, coated with PTFE, are ideal for small containers.
  • Effective stirring action
  • Suitable for food stuff