Magnetic stirring bars, PTFE, oval
供应商: KARTELL
683 684 685 686 687 688 689
KART683EA 0 CNY
KART683 KART684 KART685 KART686 KART687 KART688 KART689
Magnetic stirring bars, PTFE, oval
搅拌器 磁力搅拌子
These oval shaped PTFE stirring bars are ideal for containers or bottles with round bottom.
  • Suitable for food stuff