Spoon spatulas
供应商: KARTELL
596
KART596EA 0 CNY
KART596
Spoon spatulas
样品勺
Polyamide, fibreglass reinforced, red.
  • Good chemical resistance
  • Heat resistance up to +100 °C (up to +180 °C for short periods)
  • Robust, flexible
Certifications: DIN 12890