Jars, wide mouth, with screw cap
供应商: KARTELL
1563 1564 1565 1566 1567 1568 1569
KART1563EA 0 CNY
KART1563 KART1564 KART1565 KART1566 KART1567 KART1568 KART1569
Jars, wide mouth, with screw cap
通用罐
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from –50 to +80 °C
  • Wide opening makes it ideally suited for storing and transporting powders and pastes
With black screw cap and sealing element.
Certifications: Suitable for foodstuff according to EC regulation 10/2011.