Volumetric flasks, class B
供应商: KARTELL
1484 1485 1486 1487 1488 1489
KART1484EA 0 CNY
KART1484 KART1485 KART1486 KART1487 KART1488 KART1489
Volumetric flasks, class B
烧瓶 容量瓶
PP, translucent, with PE stopper.
  • Narrow neck with no meniscus for easy reading
  • Calibrated gravimetrically at +20 °C
  • Autoclavable, chemically non absorbent, non wetting surface