Shakers, roller 6 and 10 (basic and digital)
供应商: IKA JANKE KUNKEL
4011000
IKAA4011000EA 0 CNY
IKAA4011000
Shakers, roller 6 and 10 (basic and digital)
振荡器 旋转/ 摇摆式振荡器
Roller shakers with 6 or 10 rolls, provide smooth rocking and rolling action, basic models have fixed speed, and the digital units feature infinitely adjustable speed. Used for mixing blood samples, solid and liquid suspensions and viscous samples. The unit can be used with all conventional tubes and cylindrical bottles. Easily removable attachments enable quick cleaning in case of sample spills.
  • Can be used at 4 to 50 °C in an incubator or cold room
  • Suitable for continuous operation
  • Removable rolls enable shaking of larger tubes
  • Easy operation via touch
More Product Information
现在订购
规格
型号 Roller 6 digital
Environmentally Preferable
摇动 Rocking and rolling
速度范围 5 - 80 min⁻¹
W×D×H 240×545×115 mm
Weight 4,5 kg
最大负载 2 kg
振幅 24,5 mm