Microfine grinder, MF 10 basic
供应商: IKA JANKE KUNKEL
2870900 2871000
IKAA2870900EA 0 CNY
IKAA2870900 IKAA2871000
Microfine grinder, MF 10 basic
研磨机
Continuously operating universal grinder with powerful drive. When using the grinding heads, the processed material passes through an interchangeable stainless steel sieve. The ground material can be collected using a vessel with NS 29 standard ground joint.
  • Easy to clean stainless steel working surface
  • Choice of two different heads that can be used with the drive
  • Heads are easily interchanged
IP protection class according to DIN EN 60529: IP 22
Certifications: IP protection class according to DIN EN 60529: IP 22
订购须知 Supplied withoutMore Product Information
现在订购
规格
Environmentally Preferable
Max. size of feed granules
圆周速度
负载循环打开/关闭
电机额定输入/输出功率
过载保护
速度范围
宽度×深度×高度
重量