Dispersing elements for homogenisers
供应商: IKA JANKE KUNKEL
0593400 0594000 1024200 1713300 1713800 2348000 2404000 2466900 8003000 8003300 8003900 8005100 8006300 L004639 L004640 3304000 3305500 3370100 8039500 4447500 4447300 4447100
IKAA0593400EA 0 CNY
IKAA0593400 IKAA0594000 IKAA1024200 IKAA1713300 IKAA1713800 IKAA2348000 IKAA2404000 IKAA2466900 IKAA8003000 IKAA8003300 IKAA8003900 IKAA8005100 IKAA8006300 IKAAL004639 IKAAL004640 IKAA3304000 IKAA3305500 IKAA3370100 IKAA8039500 IKAA4447500 IKAA4447300 IKAA4447100
Dispersing elements for homogenisers
均质器 分散工具
Type N: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by all methods, max. temperature 180 °C.
Type NK: Materials in contact with medium are PTFE and stainless steel AISI 316L, suitable for solvents and abrasive substances, can be sterilised by chemical methods, max. temperature 120 °C.
Type KV: Materials in contact with medium are FFPM/SIC and stainless steel AISI 316L, suitable for vacuum down to 1 mbar, pressure up to 6 bar and for solvents, can be sterilised by chemical methods, max.More Product Information