Capillary columns, TraceGOLD™ TG-1701MS
供应商: Thermo Fisher Scientific
26090-1420
HYPE26090-1420EA 0 CNY
HYPE26090-1420
Capillary columns, TraceGOLD™ TG-1701MS
色谱柱
These columns feature a mix of cyano and phenyl groups for increased polarity and a different elution order relative to less polar columns.
  • Mid-polarity phase, 14% cyanopropylphenyl, 86% dimethyl polysiloxane
  • Fully characterised for long-term reproducibility, column-to-column consistency and low bleed
  • Optimal for confirmation analysis
  • Equivalent to USP G46 phase