Anion exchange HPLC columns, ProPac™ SAX-10 and WAX-10
供应商: Thermo Fisher Scientific
054997
HYPE054997EA 0 CNY
HYPE054997
Anion exchange HPLC columns, ProPac™ SAX-10 and WAX-10
色谱柱
These columns can resolve protein isoforms that differ by a single charged residue. A thin, hydrophilic layer prevents unwanted secondary interactions. The grafted anion exchange surface provides pH based selectivity control and fast mass transfer for high efficiency separation and moderate capacity.
  • High efficiency peaks
  • Higher capacity than other columns based on nonporous particles
  • Ideal for characterisation and quality control assessment of closely-related protein variants