碳酸氢钠 99,7 ACS, Fluka™

供应商: Honeywell Chemicals

Synonyms: Sodium bicarbonate

S6014-500G
HONCS6014-500GEA 0 CNY
HONCS6014-500G
碳酸氢钠 99,7 ACS, Fluka™
碳酸氢钠
分子式: NaHCO₃
分子量: 84.01 g/mol
溶点: 270 Dec.
密度: 2.2 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00003528
CAS号: 144-55-8
EINECS: 205-633-8
Merck Index: 13,08655
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

现在订购

产品技术规格

Identity (IR) complying
Assay (excluding dried substance) 99.7 - 100.3 %
Water insoluble matter Max. 0.015 %
Calcium (Ca) Max. 0.02 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 0.003 %
Phosphate (PO₄) Max. 0.001 %
S-compounds (as SO₄) Max. 0.003 %
Ammonium (NH₄) Max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR