Acetonitrile : Water (50:50) %, LabReady™, Solvent Blend LC-MS, HPLC, Burdick & Jackson™

供应商: Honeywell Chemicals
Danger

LC435-4
HONCLC435-4EA 0 CNY
HONCLC435-4
Acetonitrile : Water (50:50) %, LabReady™, Solvent Blend LC-MS, HPLC, Burdick & Jackson™
Acetonitrile : Water

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

闪点: 3 °C
存储条件: Ambient
EINECS: 203-466-5
UN号: 1648
ADR: 3 II

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR