Burettes, straight stopcock
供应商: HIRSCHMANN
3130160 3130170 3130175
HIRS3130160EA 0 CNY
HIRS3130160 HIRS3130170 HIRS3130175
Burettes, straight stopcock
滴定管
DURAN®, borosilicate glass, class AS.
  • Calibrated to deliver 'Ex', conformity certified (DE-M marking), with dated batch identification
  • Ring marks at major graduations, black graduations
  • With glass key
Scale length: 500 mm.
Scale length: 500 mm
Certifications: DIN ISO 385.