Bulb pipettes, one mark, Class A
供应商: HIRSCHMANN
134G1S 134G15
HIRS134G1SEA 0 CNY
HIRS134G1S HIRS134G15
Bulb pipettes, one mark, Class A
移液管和移液器 球管
Borosilicate glass.
  • Designed to deliver according to ASTM Specification E969, Class A requirements
  • Colour-coded for ease in sorting and selection
  • Fine black enamel capacity line and markings
  • Serialised pipettes include a Certificate of Identification and Accuracy
Certifications: ASTM E969