Zorbax C8, HPLC Columns
供应商: HICHROM
HICHZC8-150AEA
8458
CNY
HICHZC8-150A
HICHZC8-150AS
HICHZC8-250A
HICHZC8-10C5
HICHZC8-150AF
HICHZC8-200A
HICHZC8-250AF
Zorbax C8, HPLC Columns
色谱柱
规格
Environmentally Preferable | |
碳负载 | 12% |
加端盖的 | 是 |
表面积 | 330 m²/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...