Avantor® Hichrom, Static Mixing Tee
供应商: HICHROM
HICHHI-454EA 0 CNY
HICHHI-454
Avantor® Hichrom, Static Mixing Tee
接头
These static mixing tee are ideal for microbore or analytical gradient HPLC. It has a low swept volume (2,2 µl) and is designed for flow rates of 0,5 to 3 ml/min.
  • PEEK body with 2-piece finger tight fittings
  • Low swept volume
  • Pressure rated to 345 bar