Avantor® Hichrom, Column Frits
供应商: HICHROM
HICHHI-120EA 0 CNY
HICHHI-120 HICHHI-100 HICHHI-104 HICHHI-105 HICHHI-126 HICHHI-133
Avantor® Hichrom, Column Frits
过滤器 柱前过滤器和熔块
Frit made of 316 grade stainless steel.
  • Helps in controlling porosity
A 2 μm porosity frit is recommended for retention of 5 to 20 μm packing material and 0,5 μm porosity frit is recommended for 3 to 4 μm packing materials.
Stainless steel with encapsulated PEEK used to prevent sample and eluent from entering dead volume space between the end fitting and also with the top of the column. This results in improved peak symmetry with 1,0 to 3,2 mm Int.Ø columns.