Avantor® Hichrom, HPLC Columns, RPB
供应商: HICHROM
HICHHI-10RPB-100PEA 24806 CNY
HICHHI-10RPB-100P HICHHI-10RPB-150P HICHHI-10RPB-250P HICHHI-10RPB-50P HICHHI10RPB100SP1 HICHHI10RPB150SP1 HICHHI10RPB250SP1 HICHHIRPB-100A HICHHIRPB-100AS HICHHIRPB-100P HICHHIRPB-150A HICHHIRPB-150AS HICHHIRPB-150P HICHHIRPB-250A HICHHIRPB-250AM HICHHIRPB-250AS HICHHIRPB-250P HICHHIRPB-250SP HICHHIRPB-50P HICHHI-10RPB-100P3 HICHHI-10RPB-10CP3 HICHHI-10RPB-150P3 HICHHI-10RPB-250P3 HICHHI-10RPB-50P3 HICHHI-10RPB-50SP1 HICHHI-3.5RPB100AF HICHHI-3.5RPB100AS HICHHI-3.5RPB150AM HICHHI-3.5RPB150AS HICHHIRPB-100P3 HICHHIRPB-10C5 HICHHIRPB-150P3 HICHHIRPB-250P3 HICHHIRPB-50P3 HICHHI3.5RPB10CM5 HICHHIRPB-10C5-SK
Avantor® Hichrom, HPLC Columns, RPB
色谱柱
Avantor® Hichrom RPB is a high purity base deactivated silica which offers a unique selectivity due to the proprietary C8/C18 multi-alkyl bonding and exhaustive endcapping employed. The range includes 3,5, 5 and 10 μm phases, which are suitable for LC-MS applications and preparative separations respectively.
  • Increased productivity due to reduced analysis time
  • Decreased column length without sacrificing efficiency
  • Excellent chromatography
Avantor® Hichrom RPB is specifically designed for the reversed-phase separation of basic molecules. Residual acidicMore Product Information