Avantor® Hichrom PEEKsil™, Tubing
供应商: HICHROM
HICHHI-1083EA 0 CNY
HICHHI-1083
Avantor® Hichrom PEEKsil™, Tubing
胶管 Chromatography Tubing
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.