Vydac 214TP C4, HPLC columns
供应商: HICHROM
HICH214TP54EA 8828 CNY
HICH214TP54 HICH214FSK510 HICH214GCC510 HICH214TP5205 HICH214TP5405 HICH214TP5210 HICH214TP5410 HICH214TP5115 HICH214TP5215 HICH214TP5315 HICH214TP5415 HICH214TP51 HICH214TP52 HICH214TP53 HICH214TP510 HICH214TP10415 HICH214TP101015 HICH214TP104 HICH214TP1010 HICH214TP1022 HICH214GCC1210 HICH214FSK1210 HICH214FSK1510 HICH214TP10152215 HICH214TP101510 HICH214TP101522 HICH214GCC152010 HICH214TP152022 HICH214GK104/N HICH214GK52/N HICH214GK54/N HICH214GD104/N HICH214GD34/N HICH214GD52/N HICH214GD54/N HICH214GCC1520/N HICH214GCC1010 HICH5138110 HICH214GD51/N HICH214GK51/N
Vydac 214TP C4, HPLC columns
色谱柱
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
214TP C4 is a first generation C4 phase and is recommended for the analysis of glycoproteins,More Product Information
现在订购
规格
Environmentally Preferable
碳负载 3%
加端盖的
美国药典代码 L26