Marco Cells for Absorption Measurements with PTFE Lids, Hellma Analytics
供应商: HELLMA
6030-20-10 6030-40-10 6030-50-10
HELL6030-20-10EA 0 CNY
HELL6030-20-10 HELL6030-40-10 HELL6030-50-10
Marco Cells for Absorption Measurements with PTFE Lids, Hellma Analytics
比色皿 分光光度计比色皿
Hellma® macro cells for absorption measurements in the Vis range, 10 mm path length, with lid.
  • Cell type 6030-OG, from optical glass
  • Wavelength range from 260 to 2500 nm
  • 10 to 50 mm path length
  • With PTFE lid
  • Volume from 3,500 - 17,500 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers. Highest precision: Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.