Powder funnels
供应商: GLASWARENFABRIK KARL HECHT
44002044 44002030
HECH44002044EA 0 CNY
HECH44002044 HECH44002030
Powder funnels
漏斗 粉末漏斗
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable