Rack for centrifuge tubes
供应商: HEATHROW
HS24306
HEATHS24306EA 0 CNY
HEATHS24306
Rack for centrifuge tubes
架子 Tube Racks
ABS
  • Designed for stable use in water baths
  • Supports conical bottom centrifuge tubes
  • Alphanumeric grid reference aids tube identification
  • Sterilisation with 70% alcohol spray
For ABS plastic we do not recommend exceeding 80 °C.