96-Well deep well blocks
供应商: HAMILTON ROBOTICS
6471-01 235656
HAMR6471-01EA 0 CNY
HAMR6471-01 HAMR235656
96-Well deep well blocks
微孔板 深孔微孔板
These 96-well deep well blocks are ideal for automated processing or handling large quantities of backup samples. PP blocks are particularly suitable for backup storage of pharmaceutical substances, DNA isolation, longterm sample storage, enzyme and tissue culture applications. PS blocks are ideal for longterm storage of blood samples.
  • PP blocks are solvent resistant
  • PS blocks optically clear
  • Biological purity tested: DNase- and RNase-free, pyrogen-free (endotoxin-free), PCR inhibitor-free